Assay Details
Gene Symbol: Mrps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026080.2 | NP_080356.1 | 3 - 4 | 259 | 81 | |
GenBank mRNA | AK003117.1 | - | 3 - 4 | 243 | 81 | |
AK007338.1 | - | 3 - 4 | 243 | 81 | ||
AK010590.1 | - | 3 - 4 | 240 | 81 | ||
AK012461.1 | - | 3 - 4 | 248 | 81 | ||
AK014099.1 | - | 3 - 4 | 246 | 81 | ||
BC029262.1 | - | 3 - 4 | 233 | 4911699 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27564593 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27564590 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27564592 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | AK014099.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | AK215321.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | BC029262.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | AK012461.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | NM_026080.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | AK014099.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | AK215321.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | BC029262.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | AK012461.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507310 | NM_026080.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644309 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434693 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72307 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138543 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | AK014099.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | AK215321.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | BC029262.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | AK012461.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | NM_026080.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | AK014099.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | AK215321.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | BC029262.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | AK012461.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507310 | NM_026080.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564593 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564588 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564614 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564594 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564583 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564585 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564589 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564582 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564591 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564604 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564608 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564599 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27564611 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138543 | - | - |
Whole Mouse Genome G4122A | A_52_P507310 | AK014099.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | AK215321.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | BC029262.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | AK012461.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | NM_026080.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | AK014099.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | AK215321.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | BC029262.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | AK012461.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P507310 | NM_026080.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132869 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107847 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138543 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09168065 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09329577 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09168068 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27564590 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27564592 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27564593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27564562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564626 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09035597 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09123447 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09314134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09052184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09168068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09155197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564607 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09453566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564608 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564615 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27564564 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10384002 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384000 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384001 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384003 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438563_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450865_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10383999 | - | - |
Mouse Genome 430 2.0 Array | 1450865_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1438563_s_at | - | - |
Mouse Genome 430 2.0 Array | 1450865_s_at | - | - |
Mouse Genome 430 2.0 Array | 1438563_PM_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64660_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64660_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100020844.mm.1 | NM_026080.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020844.mm.1 | NM_026080.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020844.mm.1 | TC1100004287.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100020848.mm.1 | NM_026080.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020848.mm.1 | NM_026080.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020848.mm.1 | TC1100004287.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100020849.mm.1 | NM_026080.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020849.mm.1 | NM_026080.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020849.mm.1 | TC1100004287.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100040877.mm.1 | NM_026080.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100040877.mm.1 | NM_026080.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100040877.mm.1 | TC1100004287.mm.1 | MTA TC |
Mouse Expression Set 430 | 1450865_PM_s_at | - | - |
Mouse Expression Set 430 | 1438563_s_at | - | - |
Mouse Expression Set 430 | 1450865_s_at | - | - |
Mouse Expression Set 430 | 1438563_PM_s_at | - | - |
Murine Genome U74v2 Set | 101085_at | - | - |
Murine Genome U74v2 Set | 101085_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1438563_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1438563_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1450865_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1450865_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450865_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450865_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438563_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438563_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 907386 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5340022 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5340022 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005340022 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2673024 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2673024 | - | - |
mouse-6 v1.1 (Illumina) | scl064660.1_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5340022 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl064660.1_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340022 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2673024 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2673024 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl064660.1_30-S | - | - |
