Assay Details
| Gene Symbol: Disp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278218.1 | NP_001265147.1 | 6 - 7 | 1136 | 93 | |
| NM_001278219.1 | NP_001265148.1 | 6 - 7 | 1096 | 93 | ||
| NM_001278220.1 | NP_001265149.1 | 5 - 6 | 979 | 93 | ||
| NM_026866.3 | NP_081142.3 | 7 - 8 | 1224 | 93 | ||
| GenBank mRNA | AK004521.1 | - | 5 - 6 | 954 | 93 | |
| AK138276.1 | - | 7 - 8 | 1181 | 93 | ||
| AK170113.1 | - | 6 - 7 | 1091 | 93 | ||
| AY144589.1 | - | 5 - 6 | 934 | 93 | ||
| AY150577.1 | - | 5 - 6 | 883 | 93 | ||
| AY150698.1 | - | 7 - 8 | 1116 | 93 | ||
| BC043102.1 | - | 6 - 7 | 1097 | 6406376 | 93 | |
| BC059225.1 | - | 6 - 7 | 1097 | 6406376 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Disp1
Genomic Map