Assay Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027727.2 | NP_082003.1 | 7 - 8 | 884 | 107 | |
| GenBank mRNA | AK016957.1 | - | 7 - 8 | 812 | 107 | |
| AK049909.1 | - | 7 - 8 | 849 | 107 | ||
| AK080149.1 | - | 7 - 8 | 861 | 107 | ||
| AK081242.1 | - | 7 - 8 | 876 | 107 | ||
| AK149628.1 | - | 7 - 8 | 858 | 107 | ||
| AK149879.1 | - | 7 - 8 | 859 | 107 | ||
| AK149891.1 | - | 7 - 8 | 858 | 107 | ||
| AK154794.1 | - | 7 - 8 | 873 | 107 | ||
| BC018220.1 | - | 3 - 4 | 244 | 3985017 | 107 | |
| BC038276.1 | - | 7 - 8 | 886 | 4458792 | 107 | |
More Information
Set Membership: |
|

Dym
Genomic Map