Assay Details
| Gene Symbol: Prkcdbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028444.1 | NP_082720.1 | 1 - 2 | 419 | 120 | |
| GenBank mRNA | AK014051.1 | - | 1 - 2 | 150 | 120 | |
| AK032154.1 | - | 1 - 2 | 199 | 120 | ||
| BC009660.1 | - | 1 - 2 | 419 | 3486461 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392593 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392590 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392588 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392591 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392587 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P180974 | AK014051.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P180974 | AK032154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P180974 | NM_028444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P180974 | BC009660.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P180974 | AK014051.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P180974 | AK032154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P180974 | NM_028444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P180974 | BC009660.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484000 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168592 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136170 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P180974 | AK014051.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P180974 | AK032154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P180974 | NM_028444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P180974 | BC009660.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P180974 | AK014051.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P180974 | AK032154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P180974 | NM_028444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P180974 | BC009660.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25392614 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168592 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136170 | - | - |
| Whole Mouse Genome G4122A | A_52_P1164479 | AK048894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180974 | AK014051.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180974 | AK032154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180974 | NM_028444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180974 | BC009660.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180974 | AK014051.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180974 | AK032154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180974 | NM_028444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180974 | BC009660.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110963 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136170 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168592 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262485 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262487 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262514 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262494 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262507 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392587 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392591 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392588 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392593 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25392590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25392569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06152811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06458596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05986620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06171876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06145078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06594578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06262503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06458589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06262507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25392575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979526 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566456 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423771_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566454 | - | - |
| Mouse Genome 430 2.0 Array | 1423771_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423771_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109042_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109042_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700033104.mm.1 | NM_028444.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033104.mm.1 | NM_028444.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033104.mm.1 | TC0700003978.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700033103.mm.1 | NM_028444.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033103.mm.1 | NM_028444.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033103.mm.1 | TC0700003978.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423771_at | - | - |
| Mouse Expression Set 430 | 1423771_PM_at | - | - |
| Murine Genome U74v2 Set | 97496_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97496_f_at | - | - |
| Murine Genome U74v2 Set | 170793_at | - | - |
| Murine Genome U74v2 Set | 170793_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423771_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423771_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423771_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423771_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 926361 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4210341 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210341 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243602 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243602 | - | - |
| mouse-6 v1.1 (Illumina) | scl30887.1.106_97-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30887.1.106_97-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210341 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1243602 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2956942 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2956942 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243602 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl30887.1.106_97-S | - | - |

Prkcdbp
Genomic Map