Assay Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028731.5 | NP_083007.2 | 12 - 13 | 1445 | 56 | |
| GenBank mRNA | AK014813.1 | - | 12 - 13 | 1446 | 56 | |
| AK044360.1 | - | 12 - 13 | 1447 | 56 | ||
| AK157352.1 | - | 12 - 13 | 1414 | 56 | ||
| BC040392.1 | - | 1 - 2 | 65 | 56 | ||
| BC052440.1 | - | 2 - 3 | 136 | 56 | ||
| BC059230.1 | - | 2 - 3 | 136 | 56 | ||
| BC138937.1 | - | 12 - 13 | 1354 | 8861959 | 56 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK014813.1,AK044360.1,BC052440.1,BC059230.1 transcripts.
Set Membership: |
|

Esyt2
Genomic Map