Assay Details
| Gene Symbol: Raf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029780.3 | NP_084056.1 | 9 - 10 | 1422 | 99 | |
| GenBank mRNA | AB057655.1 | - | 8 - 9 | 967 | 99 | |
| AB057663.2 | - | 9 - 10 | 1192 | 99 | ||
| AK141745.1 | - | 9 - 10 | 1423 | 99 | ||
| AK148327.1 | - | 331 | 99 | |||
| BC015273.1 | - | 9 - 10 | 1301 | 4222659 | 99 | |
| BC092040.1 | - | 9 - 10 | 1287 | 2645313 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Raf1
Genomic Map