Assay Details
| Gene Symbol: Socs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080843.2 | NP_543119.2 | 2 | 915 | 73 | |
| GenBank mRNA | AK014115.1 | - | 2 | 946 | 73 | |
| AW046781.1 | - | 1 | 330 | 73 | ||
| AY046322.1 | - | 1 | 743 | 73 | ||
| BC117814.1 | - | 1 | 836 | 40092681 | 73 | |
| BC117815.1 | - | 1 | 836 | 40092683 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein kinase activity
negative regulation of epidermal growth factor-activated receptor activity
negative regulation of signal transduction
cytokine-mediated signaling pathway
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
regulation of growth
negative regulation of epidermal growth factor receptor signaling pathway
negative regulation of JAK-STAT cascade
molecular_function
protein kinase inhibitor activity
negative regulation of protein kinase activity
negative regulation of epidermal growth factor-activated receptor activity
negative regulation of signal transduction
cytokine-mediated signaling pathway
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
regulation of growth
negative regulation of epidermal growth factor receptor signaling pathway
negative regulation of JAK-STAT cascade
molecular_function
protein kinase inhibitor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29581356 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29581357 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973886 | BC117814.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973886 | BC117815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973886 | BG083279.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973886 | NM_080843.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973886 | AK014988.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110309 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499168 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12636 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154181 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149567 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973886 | BC117814.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973886 | BC117815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973886 | BG083279.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973886 | NM_080843.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973886 | AK014988.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29581369 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149567 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08102013 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08102020 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154181 | - | - |
| Whole Mouse Genome G4122A | A_51_P165424 | BC117814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165424 | BC117815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165424 | AY046322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165424 | NM_080843.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165424 | AK014115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P212065 | BC117814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P212065 | BC117815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P212065 | AW046781.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P212065 | AY046322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P212065 | BC057159.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126017 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149567 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11752826 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11752836 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11752820 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11752838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29581357 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29581356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29581471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11423335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11636632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11658277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11786673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11756571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29581425 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10414352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414354 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421274_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455142_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421273_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421275_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10414350 | - | - |
| Mouse Genome 430 2.0 Array | 1421275_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1421274_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421273_at | - | - |
| Mouse Genome 430 2.0 Array | 1421273_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455142_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455142_at | - | - |
| Mouse Genome 430 2.0 Array | 1421274_at | - | - |
| Mouse Genome 430 2.0 Array | 1421275_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67296_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67296_PM_at | - | - |
| Mouse Expression Set 430 | 1421274_at | - | - |
| Mouse Expression Set 430 | 1421273_at | - | - |
| Mouse Expression Set 430 | 1421274_PM_at | - | - |
| Mouse Expression Set 430 | 1421275_PM_s_at | - | - |
| Mouse Expression Set 430 | 1421273_PM_at | - | - |
| Mouse Expression Set 430 | 1421275_s_at | - | - |
| Mouse Expression Set 430 | 1455142_at | - | - |
| Mouse Expression Set 430 | 1455142_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421274_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421273_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421275_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421273_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421275_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421274_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421274_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421274_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421275_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421273_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421275_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421273_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455142_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455142_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 716324 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1170162 | - | - |
| MouseWG-6 v2 Expression BeadChip | 101850064 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101850064 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1170162 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170162 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249426 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240921 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249426 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067296.2_190-S | - | - |
| mouse-6 v1.1 (Illumina) | scl46405.2_247-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170162 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101850064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067296.2_190-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170162 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101850064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1249426 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249426 | - | - |

Socs4
Genomic Map