Assay Details
| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080849.3 | NP_543125.1 | 8 - 9 | 1299 | 83 | |
| GenBank mRNA | AF407579.1 | - | 8 - 9 | 1249 | 83 | |
| AK014546.1 | - | 6 - 7 | 1475 | 83 | ||
| AK154358.1 | - | 8 - 9 | 1262 | 83 | ||
| BC057995.1 | - | 7 - 8 | 1381 | 83 | ||
| BC070457.1 | - | 8 - 9 | 1248 | 30603080 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek8
Genomic Map