Assay Details
| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080849.3 | NP_543125.1 | 10 - 11 | 1495 | 81 | |
| GenBank mRNA | AF407579.1 | - | 10 - 11 | 1445 | 81 | |
| AK014546.1 | - | 8 - 9 | 1671 | 81 | ||
| AK154358.1 | - | 10 - 11 | 1458 | 81 | ||
| BC057995.1 | - | 9 - 10 | 1577 | 81 | ||
| BC070457.1 | - | 10 - 11 | 1444 | 30603080 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek8
Genomic Map