Assay Details
| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080849.3 | NP_543125.1 | 11 - 12 | 1642 | 104 | |
| GenBank mRNA | AF407579.1 | - | 11 - 12 | 1592 | 104 | |
| AK014546.1 | - | 9 - 10 | 1818 | 104 | ||
| BC057995.1 | - | 10 - 11 | 1724 | 104 | ||
| BC070457.1 | - | 11 - 12 | 1591 | 30603080 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek8
Genomic Map