Assay Details
| Gene Symbol: Sult1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133670.1 | NP_598431.1 | 6 - 7 | 639 | 87 | |
| GenBank mRNA | AK002700.1 | - | 6 - 7 | 662 | 87 | |
| BC005413.1 | - | 6 - 7 | 639 | 3488459 | 87 | |
| L02331.1 | - | 6 - 7 | 655 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
catecholamine metabolic process
lipid metabolic process
xenobiotic metabolic process
steroid metabolic process
estrogen metabolic process
drug metabolic process
4-nitrophenol metabolic process
response to glucocorticoid
sulfation
nucleotide binding
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
aryl sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
tyrosine-ester sulfotransferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
protein homodimerization activity
3'-phosphoadenosine 5'-phosphosulfate binding
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
catecholamine metabolic process
lipid metabolic process
xenobiotic metabolic process
steroid metabolic process
estrogen metabolic process
drug metabolic process
4-nitrophenol metabolic process
response to glucocorticoid
sulfation
nucleotide binding
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
aryl sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
tyrosine-ester sulfotransferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
protein homodimerization activity
3'-phosphoadenosine 5'-phosphosulfate binding
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005475 | NM_133670.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005475 | AK002700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005475 | BC005413.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321341 | NM_133670.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321341 | AK002700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321341 | BC005413.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321341 | NM_133670.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321341 | AK002700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321341 | BC005413.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606385 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419410 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76371 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139677 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321341 | NM_133670.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321341 | AK002700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321341 | BC005413.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321341 | NM_133670.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321341 | AK002700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321341 | BC005413.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005475 | NM_133670.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005475 | AK002700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005475 | BC005413.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321341 | NM_133670.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321341 | AK002700.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321341 | BC005413.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321341 | NM_133670.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321341 | AK002700.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321341 | BC005413.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501686 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139677 | - | - |
| Whole Mouse Genome G4122A | A_51_P321341 | NM_133670.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321341 | AK002700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321341 | BC005413.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321341 | NM_133670.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321341 | AK002700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321341 | BC005413.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114313 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139677 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299328 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299363 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299364 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299332 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299362 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299349 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299346 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05967225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05967227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06651008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05942515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06252238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05967221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06299333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06648684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568002 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427345_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568001 | - | - |
| Mouse Genome 430 2.0 Array | 1427345_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427345_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20887_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20887_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700035856.mm.1 | NM_133670.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035856.mm.1 | NM_133670.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035856.mm.1 | TC0700004291.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700035861.mm.1 | NM_133670.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035861.mm.1 | NM_133670.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035861.mm.1 | TC0700004291.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700035857.mm.1 | NM_133670.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035857.mm.1 | NM_133670.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035857.mm.1 | TC0700004291.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700035858.mm.1 | NM_133670.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035858.mm.1 | NM_133670.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035858.mm.1 | TC0700004291.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700035859.mm.1 | NM_133670.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035859.mm.1 | NM_133670.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035859.mm.1 | TC0700004291.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 162130_r_at | - | - |
| Murine Genome U74v2 Set | 162130_PM_r_at | - | - |
| Murine Genome U74v2 Set | 103087_PM_at | - | - |
| Murine Genome U74v2 Set | 103087_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427345_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427345_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 619584 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5130706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103830450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2745370 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2745370 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234145 | - | - |
| mouse-6 v1.1 (Illumina) | scl000152.1_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl30678.8_112-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103830450 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30678.8_112-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130706 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103830450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2745370 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2745370 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl30678.8_112-S | - | - |

Sult1a1
Genomic Map