Assay Details
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133701.2 | NP_598462.1 | 6 - 7 | 913 | 67 | |
GenBank mRNA | AK081998.1 | - | 6 - 7 | 848 | 67 | |
AK145461.1 | - | 6 - 7 | 916 | 67 | ||
AK166890.1 | - | 6 - 7 | 865 | 67 | ||
BC014869.1 | - | 6 - 7 | 840 | 3602158 | 67 | |
BC023691.1 | - | 6 - 7 | 842 | 5344332 | 67 | |
BC025030.1 | - | 6 - 7 | 859 | 4947416 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
