Assay Details
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133701.2 | NP_598462.1 | 8 - 9 | 1169 | 112 | |
GenBank mRNA | AK081998.1 | - | 8 - 9 | 1104 | 112 | |
AK145461.1 | - | 8 - 9 | 1172 | 112 | ||
AK166890.1 | - | 8 - 9 | 1121 | 112 | ||
AK178683.1 | - | 1 - 2 | 174 | 113 | ||
AK189258.1 | - | 1 - 2 | 173 | 112 | ||
AK219394.1 | - | 1 - 2 | 174 | 112 | ||
BC014869.1 | - | 8 - 9 | 1096 | 3602158 | 112 | |
BC023691.1 | - | 8 - 9 | 1098 | 5344332 | 112 | |
BC025030.1 | - | 8 - 9 | 1115 | 4947416 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
