Assay Details
| Gene Symbol: Rdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133832.3 | NP_598593.1 | 4 - 5 | 902 | 87 | |
| GenBank mRNA | AF456767.1 | - | 4 - 5 | 1010 | 87 | |
| AK149656.1 | - | 4 - 5 | 973 | 87 | ||
| BC019796.1 | - | 4 - 5 | 902 | 4923005 | 87 | |
| BC038334.1 | - | 2 - 3 | 87 | |||
| BC052546.1 | - | 2 - 3 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rdh10
Genomic Map