Assay Details
| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177992.2 | NP_818773.1 | 5 - 6 | 630 | 121 | |
| GenBank mRNA | AK031004.1 | - | 5 - 6 | 632 | 121 | |
| AK075772.1 | - | 5 - 6 | 588 | 121 | ||
| BC003886.1 | - | 5 - 6 | 589 | 3590411 | 121 | |
| BC024109.1 | - | 5 - 6 | 586 | 5149353 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | BC003886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | BC024109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | AK031004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | AK075772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | NM_177992.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | BC003886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | BC024109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | AK031004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | AK075772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430327 | NM_177992.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680520 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140987 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | BC003886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | BC024109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | AK031004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | AK075772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | NM_177992.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | BC003886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | BC024109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | AK031004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | AK075772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430327 | NM_177992.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | BC003886.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | BC024109.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | AK031004.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | AK075772.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | NM_177992.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | BC003886.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | BC024109.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | AK031004.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | AK075772.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430327 | NM_177992.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166467 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08119220 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08119212 | - | - |
| Whole Mouse Genome G4122A | A_51_P430327 | BC003886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | BC024109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | AK031004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | AK075772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | NM_177992.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | BC003886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | BC024109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | AK031004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | AK075772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430327 | NM_177992.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11784145 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11195324 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11784143 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11784149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11129948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11768146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11195340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11129951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11784145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11129949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29619222 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10415371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415368 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415369 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415372 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429180_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416356_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415363 | - | - |
| Mouse Genome 430 2.0 Array | 1429180_at | - | - |
| Mouse Genome 430 2.0 Array | 1416356_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429180_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416356_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105446_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105446_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400006418.mm.1 | NM_177992.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006418.mm.1 | NM_177992.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006418.mm.1 | TC1400000790.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400006419.mm.1 | NM_177992.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006419.mm.1 | NM_177992.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006419.mm.1 | TC1400000790.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416356_at | - | - |
| Mouse Expression Set 430 | 1416356_PM_at | - | - |
| Mouse Expression Set 430 | 1429180_at | - | - |
| Mouse Expression Set 430 | 1429180_PM_at | - | - |
| Murine Genome U74v2 Set | 139141_PM_s_at | - | - |
| Murine Genome U74v2 Set | 139142_r_at | - | - |
| Murine Genome U74v2 Set | 139142_PM_r_at | - | - |
| Murine Genome U74v2 Set | 139141_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416356_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416356_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416356_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416356_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429180_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429180_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4920204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4920204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2617691 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2617691 | - | - |
| mouse-6 v1.1 (Illumina) | scl000307.1_98-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920204 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000307.1_98-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920204 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2846317 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2617691 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2846317 | - | - |

Gmpr2
Genomic Map