Assay Details
| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177992.2 | NP_818773.1 | 7 - 8 | 817 | 87 | |
| GenBank mRNA | AK031004.1 | - | 7 - 8 | 819 | 87 | |
| AK075772.1 | - | 7 - 8 | 775 | 87 | ||
| AK180702.1 | - | 3 - 4 | 332 | 87 | ||
| BC003886.1 | - | 7 - 8 | 776 | 3590411 | 87 | |
| BC024109.1 | - | 7 - 8 | 773 | 5149353 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmpr2
Genomic Map