Assay Details
| Gene Symbol: Brms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134155.1 | NP_598916.1 | 3 - 4 | 325 | 75 | |
| GenBank mRNA | AF233580.1 | - | 3 - 4 | 336 | 75 | |
| AK076729.1 | - | 3 - 4 | 332 | 76 | ||
| BC003485.1 | - | 3 - 4 | 343 | 2811430 | 75 | |
| BC016108.1 | - | 3 - 4 | 325 | 4921658 | 75 | |
| BC038380.1 | - | 2564 | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Brms1
Genomic Map