Assay Details
| Gene Symbol: Brms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134155.1 | NP_598916.1 | 6 - 7 | 633 | 62 | |
| GenBank mRNA | AF233580.1 | - | 6 - 7 | 644 | 62 | |
| AK076729.1 | - | 6 - 7 | 641 | 62 | ||
| BC016108.1 | - | 6 - 7 | 633 | 4921658 | 62 | |
| BC038380.1 | - | 2872 | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Brms1
Genomic Map