Assay Details
| Gene Symbol: Il17re | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034029.1 | NP_001029201.1 | 11 - 12 | 1161 | 82 | |
| NM_001034031.1 | NP_001029203.1 | 13 - 14 | 1236 | 82 | ||
| NM_145826.5 | NP_665825.2 | 12 - 13 | 1588 | 82 | ||
| GenBank mRNA | AF458068.1 | - | 12 - 13 | 1238 | 82 | |
| AK046302.1 | - | 12 - 13 | 1591 | 82 | ||
| AK046713.1 | - | 12 - 13 | 1250 | 82 | ||
| AK078876.1 | - | 4 - 5 | 1502 | 82 | ||
| AK169498.1 | - | 12 - 13 | 1416 | 82 | ||
| BC069861.1 | - | 12 - 13 | 1265 | 6311568 | 82 | |
| DQ092339.1 | - | 12 - 13 | 1177 | 82 | ||
| DQ092340.1 | - | 11 - 12 | 1161 | 82 | ||
| DQ092341.1 | - | 13 - 14 | 1236 | 82 | ||
More Information
Set Membership: |
|

Il17re
Genomic Map