Assay Details
Gene Symbol: Sec14l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146013.1 | NP_666125.1 | 10 - 11 | 977 | 73 | |
GenBank mRNA | AK028954.1 | - | 9 - 10 | 1220 | 73 | |
BC024073.1 | - | 3 - 4 | 312 | 73 | ||
BC026948.1 | - | 10 - 11 | 977 | 5136664 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194853 | BC024073.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194853 | BC026948.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194853 | NM_146013.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194853 | AK028954.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194853 | BC024073.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194853 | BC026948.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194853 | NM_146013.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194853 | AK028954.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555414 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 217879 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623376 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319126 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601056 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505505 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584841 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521803 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938518 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380691 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40559 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142512 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178805 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194853 | BC024073.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194853 | BC026948.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194853 | NM_146013.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194853 | AK028954.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194853 | BC024073.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194853 | BC026948.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194853 | NM_146013.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194853 | AK028954.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194853 | BC024073.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194853 | BC026948.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194853 | NM_146013.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194853 | AK028954.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194853 | BC024073.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194853 | BC026948.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194853 | NM_146013.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194853 | AK028954.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555260 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27555241 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02251153 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07239664 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142512 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178805 | - | - |
Whole Mouse Genome G4122A | A_51_P194853 | BC024073.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P194853 | BC026948.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P194853 | NM_146013.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P194853 | AK028954.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P194853 | BC024073.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P194853 | BC026948.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P194853 | NM_146013.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P194853 | AK028954.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110150 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142512 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178805 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09135457 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09135453 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09135456 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09135454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27555318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09135453 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09369606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09135457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555236 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555279 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555282 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09660334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555258 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555241 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09410502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555289 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09410879 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09410504 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09135454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09662875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09539373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27555260 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10373837 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373845 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373840 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373839 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373843 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373841 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373835 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373838 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373844 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373836 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373842 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424676_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10373834 | - | - |
Mouse Genome 430 2.0 Array | 1424676_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1424676_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103655_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103655_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100000391.mm.1 | NM_146013.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000391.mm.1 | NM_146013.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000391.mm.1 | TC1100000032.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100000392.mm.1 | NM_146013.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000392.mm.1 | NM_146013.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000392.mm.1 | TC1100000032.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100000394.mm.1 | NM_146013.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000394.mm.1 | NM_146013.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000394.mm.1 | TC1100000032.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100000393.mm.1 | NM_146013.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000393.mm.1 | NM_146013.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100000393.mm.1 | TC1100000032.mm.1 | MTA TC |
Affymetrix HT Mouse Genome 430A Array | 1424676_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424676_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 932918 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 7040458 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 7040458 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007040458 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605013 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2605013 | - | - |
mouse-6 v1.1 (Illumina) | scl41894.14.178_7-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7040458 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl41894.14.178_7-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040458 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2605013 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2837447 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2605013 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2837447 | - | - |
