Assay Details
| Gene Symbol: Ndufs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153064.4 | NP_694704.1 | 2 - 3 | 226 | 66 | |
| GenBank mRNA | AK078474.1 | - | 2 - 3 | 227 | 66 | |
| AK150431.1 | - | 2 - 3 | 222 | 66 | ||
| AK151746.1 | - | 2 - 3 | 199 | 66 | ||
| AK153364.1 | - | 2 - 3 | 199 | 66 | ||
| AK165426.1 | - | 2 - 3 | 222 | 66 | ||
| AK167016.1 | - | 2 - 3 | 224 | 66 | ||
| AK168971.1 | - | 2 - 3 | 219 | 66 | ||
| BC003898.1 | - | 2 - 3 | 225 | 66 | ||
| BC016097.1 | - | 2 - 3 | 213 | 4910260 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs2
Genomic Map