Assay Details
| Gene Symbol: Ehd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153068.3 | NP_694708.2 | 4 - 5 | 1087 | 79 | |
| GenBank mRNA | AK029240.1 | - | 4 - 5 | 1031 | 79 | |
| AK031203.1 | - | 4 - 5 | 1089 | 79 | ||
| AK046566.1 | - | 4 - 5 | 1029 | 79 | ||
| AK048356.1 | - | 4 - 5 | 1032 | 79 | ||
| AK050243.1 | - | 4 - 5 | 1067 | 79 | ||
| AK084455.1 | - | 4 - 5 | 1030 | 79 | ||
| AK085016.1 | - | 4 - 5 | 1030 | 79 | ||
| AY531389.1 | - | 4 - 5 | 1026 | 79 | ||
| BC027084.1 | - | 5 - 6 | 734 | 5356224 | 79 | |
| BC031731.1 | - | 952 | 79 | |||
| BC113161.1 | - | 5 - 6 | 1045 | 40060184 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ehd2
Genomic Map