Assay Details
| Gene Symbol: Slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153150.2 | NP_694790.1 | 4 - 5 | 579 | 62 | |
| GenBank mRNA | AK005070.1 | - | 4 - 5 | 509 | 62 | |
| AK159289.1 | - | 4 - 5 | 542 | 62 | ||
| AK170191.1 | - | 4 - 5 | 543 | 62 | ||
| AK188281.1 | - | 1 - 2 | 47 | 62 | ||
| AK206906.1 | - | 1 - 2 | 47 | 62 | ||
| AK211700.1 | - | 1 - 2 | 47 | 62 | ||
| AK219123.1 | - | 1 - 2 | 47 | 62 | ||
| BC037087.1 | - | 4 - 5 | 539 | 5124403 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a1
Genomic Map