Assay Details
Gene Symbol: Grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153419.2 | NP_700468.2 | 5 - 6 | 834 | 79 | |
GenBank mRNA | AB001539.1 | - | 6 - 7 | 906 | 79 | |
AK077611.1 | - | 5 - 6 | 840 | 79 | ||
BC008121.1 | - | 5 - 6 | 811 | 79 | ||
BC028896.1 | - | 4 - 5 | 703 | 3674738 | 79 | |
BC083143.1 | - | 5 - 6 | 822 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
