Assay Details
Gene Symbol: Elof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK002505.1 | - | 63 | 81 | ||
AK003614.1 | - | 67 | 81 | |||
AK014116.1 | - | 43 | 81 | |||
AK089052.1 | - | 53 | 81 | |||
BC019870.1 | - | 36 | 5123727 | 81 | ||
BC024488.1 | - | 48 | 4981209 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK002505.1,AK003614.1,AK014116.1,AK089052.1,BC019870.1,BC024488.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
