Assay Details
| Gene Symbol: Elk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007922.5 | NP_031948.4 | 1 - 2 | 168 | 60 | |
| GenBank mRNA | AK082260.1 | - | 1 - 2 | 168 | 60 | |
| AK132354.1 | - | 1 - 2 | 161 | 60 | ||
| BC054474.1 | - | 1 - 2 | 152 | 30045506 | 60 | |
| X87257.1 | - | 1 - 2 | 60 | |||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK132354.1,BC054474.1,NM_007922.5,X87257.1 transcripts.
Set Membership: |
|

Elk1
Genomic Map