Assay Details
| Gene Symbol: Elk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007922.5 | NP_031948.4 | 2 - 3 | 419 | 82 | |
| GenBank mRNA | AK132354.1 | - | 2 - 3 | 412 | 82 | |
| BC054474.1 | - | 2 - 3 | 403 | 30045506 | 82 | |
| CV561936.1 | - | 1 - 2 | 365 | 82 | ||
| X87257.1 | - | 2 - 3 | 689 | 82 | ||
| Z36939.1 | - | 1 - 2 | 204 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elk1
Genomic Map