Assay Details
Gene Symbol: Enpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007934.3 | NP_031960.1 | 10 - 11 | 1973 | 116 | |
GenBank mRNA | AK031439.1 | - | 9 - 10 | 1005 | 116 | |
AK128975.1 | - | 10 - 11 | 1930 | 116 | ||
AK146808.1 | - | 10 - 11 | 1945 | 116 | ||
BC127060.1 | - | 10 - 11 | 1837 | 40109863 | 116 | |
DQ000292.1 | - | 10 - 11 | 1713 | 116 | ||
M29961.1 | - | 10 - 11 | 1792 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
