Assay Details
| Gene Symbol: Etv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007959.2 | NP_031985.2 | 2 - 3 | 185 | 85 | |
| GenBank mRNA | L10427.1 | - | 2 - 3 | 185 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etv2
Genomic Map