Assay Details
| Gene Symbol: Hcls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008225.2 | NP_032251.2 | 5 - 6 | 496 | 132 | |
| GenBank mRNA | AK088723.1 | - | 5 - 6 | 478 | 132 | |
| BC007469.1 | - | 5 - 6 | 466 | 3593456 | 132 | |
| D42120.1 | - | 5 - 6 | 427 | 132 | ||
| X84797.1 | - | 5 - 6 | 464 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hcls1
Genomic Map