Assay Details
| Gene Symbol: Hcls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008225.2 | NP_032251.2 | 10 - 11 | 990 | 64 | |
| GenBank mRNA | AK088723.1 | - | 10 - 11 | 972 | 64 | |
| AK132608.1 | - | 1 - 2 | 38 | 64 | ||
| BC007469.1 | - | 10 - 11 | 960 | 3593456 | 64 | |
| CA321698.1 | - | 1 - 2 | 72 | 64 | ||
| D42120.1 | - | 10 - 11 | 921 | 64 | ||
| X84797.1 | - | 10 - 11 | 958 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hcls1
Genomic Map