Assay Details
| Gene Symbol: Hdgfrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143616.1 | - | 1 - 2 | 187 | 95 | |
| AK144669.1 | - | 1 - 2 | 191 | 95 | ||
| AK146813.1 | - | 1 - 2 | 126 | 95 | ||
| AK146918.1 | - | 1 - 2 | 164 | 95 | ||
| BC003741.1 | - | 1 - 2 | 123 | 3488803 | 95 | |
| D63850.1 | - | 1 - 2 | 167 | 95 | ||
| FN687734.2 | - | 1 - 2 | 69 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdgfrp2
Genomic Map