Assay Details
| Gene Symbol: Hdgfrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143616.1 | - | 12 - 13 | 1578 | 68 | |
| AK144669.1 | - | 12 - 13 | 1585 | 68 | ||
| AK146813.1 | - | 12 - 13 | 1547 | 68 | ||
| AK146918.1 | - | 12 - 13 | 1555 | 68 | ||
| BC003741.1 | - | 12 - 13 | 1544 | 3488803 | 68 | |
| D63850.1 | - | 12 - 13 | 1561 | 68 | ||
| FN687734.2 | - | 12 - 13 | 1304 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdgfrp2
Genomic Map