Assay Details
| Gene Symbol: Mst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008243.3 | NP_032269.3 | 13 - 14 | 1559 | 77 | |
| GenBank mRNA | AK040934.1 | - | 13 - 14 | 1543 | 77 | |
| AK134222.1 | - | 13 - 14 | 1542 | 77 | ||
| AK144824.1 | - | 13 - 14 | 1522 | 77 | ||
| BC010551.1 | - | 13 - 14 | 1544 | 4195560 | 77 | |
| BC051393.1 | - | 13 - 14 | 1528 | 5055055 | 77 | |
| M74181.1 | - | 13 - 14 | 1516 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115392 | BC010551.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984098 | M74181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984098 | AK144824.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984098 | NM_008243.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984098 | AK134222.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818227 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161784 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984098 | M74181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984098 | AK144824.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984098 | NM_008243.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984098 | AK134222.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115392 | BC010551.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P290191 | M74181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P290191 | AK144824.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P290191 | BC051393.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P290191 | NM_008243.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P290191 | AK040934.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812869 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161784 | - | - |
| Whole Mouse Genome G4122A | A_51_P290191 | M74181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290191 | AK144824.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290191 | BC051393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290191 | NM_008243.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290191 | AK040934.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100091 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161784 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07946865 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07946811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07946808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07946843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07946836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07946813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07757223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812839 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588857 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588858 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588865 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588860 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588863 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588856 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588862 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588867 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588861 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588859 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588869 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418267_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588855 | - | - |
| Mouse Genome 430 2.0 Array | 1418267_at | - | - |
| Mouse Genome 430 2.0 Array | 1418267_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15235_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15235_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900011185.mm.1 | NM_008243.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011185.mm.1 | NM_008243.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011185.mm.1 | TC0900001393.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418267_at | - | - |
| Mouse Expression Set 430 | 1418267_PM_at | - | - |
| Murine Genome U74v2 Set | 100286_at | - | - |
| Murine Genome U74v2 Set | 100286_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418267_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418267_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418267_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418267_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1400403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1400403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2681885 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2681885 | - | - |
| mouse-6 v1.1 (Illumina) | scl015235.16_24-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl015235.16_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2847887 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2847887 | - | - |

Mst1
Genomic Map