Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100019517.mm.1 |
NM_001159328.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019517.mm.1 |
NM_001159328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019517.mm.1 |
NM_008244.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019517.mm.1 |
NM_008244.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019517.mm.1 |
TC1100002028.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100019518.mm.1 |
NM_001159328.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019518.mm.1 |
NM_001159328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019518.mm.1 |
NM_008244.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019518.mm.1 |
NM_008244.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019518.mm.1 |
TC1100002028.mm.1 |
MTA TC |