Assay Details
| Gene Symbol: Hiat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008246.2 | NP_032272.2 | 6 - 7 | 904 | 87 | |
| GenBank mRNA | AK004779.1 | - | 6 - 7 | 843 | 87 | |
| AK135865.1 | - | 6 - 7 | 725 | 87 | ||
| AK158838.1 | - | 6 - 7 | 905 | 87 | ||
| AK169626.1 | - | 6 - 7 | 844 | 87 | ||
| BC118047.1 | - | 6 - 7 | 724 | 40106779 | 87 | |
| D88315.1 | - | 6 - 7 | 777 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hiat1
Genomic Map