Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008246.2 | NP_032272.2 | 8 - 9 | 1088 | 100 | |
GenBank mRNA | AK004779.1 | - | 8 - 9 | 1027 | 100 | |
AK135865.1 | - | 8 - 9 | 909 | 100 | ||
AK158838.1 | - | 8 - 9 | 1089 | 100 | ||
AK169626.1 | - | 8 - 9 | 1028 | 100 | ||
BC118047.1 | - | 8 - 9 | 908 | 40106779 | 100 | |
D88315.1 | - | 8 - 9 | 961 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
