Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008246.2 | NP_032272.2 | 9 - 10 | 1202 | 84 | |
GenBank mRNA | AK004779.1 | - | 9 - 10 | 1141 | 84 | |
AK135865.1 | - | 9 - 10 | 1023 | 84 | ||
AK158838.1 | - | 9 - 10 | 1203 | 84 | ||
AK169626.1 | - | 9 - 10 | 1142 | 84 | ||
BC118047.1 | - | 9 - 10 | 1022 | 40106779 | 84 | |
D88315.1 | - | 9 - 10 | 1075 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
