Assay Details
| Gene Symbol: Hmgcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008254.2 | NP_032280.2 | 5 - 6 | 527 | 93 | |
| GenBank mRNA | AK132035.1 | - | 5 - 6 | 516 | 93 | |
| AK144768.1 | - | 5 - 6 | 525 | 93 | ||
| AK145251.1 | - | 5 - 6 | 523 | 93 | ||
| AK154033.1 | - | 5 - 6 | 516 | 93 | ||
| BC025440.1 | - | 5 - 6 | 514 | 5049759 | 93 | |
| S65036.1 | - | 5 - 6 | 515 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcl
Genomic Map