Assay Details
| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010163.3 | NP_034293.2 | 1 - 2 | 178 | 104 | |
| GenBank mRNA | AK084772.1 | - | 1 - 2 | 160 | 104 | |
| AK148090.1 | - | 1 - 2 | 167 | 104 | ||
| AK150155.1 | - | 1 - 2 | 170 | 104 | ||
| AK164342.1 | - | 1 - 2 | 164 | 104 | ||
| BB842500.1 | - | 1 - 2 | 179 | 104 | ||
| BC006597.1 | - | 1 - 2 | 151 | 3965079 | 104 | |
| U67837.1 | - | 1 - 2 | 153 | 104 | ||
| U72141.1 | - | 1 - 2 | 157 | 104 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK084772.1,AK164342.1,BC006597.1,NM_010163.3,U67837.1,U72141.1 transcripts.
Set Membership: |
|

Ext2
Genomic Map