Assay Details
| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010163.3 | NP_034293.2 | 9 - 10 | 1705 | 92 | |
| GenBank mRNA | AK082572.1 | - | 9 - 10 | 1687 | 92 | |
| AK134438.1 | - | 5 - 6 | 646 | 92 | ||
| AK148090.1 | - | 9 - 10 | 1695 | 92 | ||
| AK148369.1 | - | 8 - 9 | 1466 | 92 | ||
| AK150155.1 | - | 9 - 10 | 1697 | 92 | ||
| AK152265.1 | - | 1805 | 92 | |||
| AK152840.1 | - | 1807 | 92 | |||
| AK164342.1 | - | 9 - 10 | 1691 | 92 | ||
| AK218011.1 | - | 1 - 2 | 83 | 92 | ||
| BC006597.1 | - | 9 - 10 | 1678 | 3965079 | 92 | |
| U67837.1 | - | 9 - 10 | 1680 | 92 | ||
| U72141.1 | - | 9 - 10 | 1684 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ext2
Genomic Map