Assay Details
| Gene Symbol: H2-Ob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010389.3 | NP_034519.2 | 3 - 4 | 710 | 78 | |
| GenBank mRNA | AK053233.1 | - | 3 - 4 | 759 | 78 | |
| KJ650230.1 | - | 3 - 4 | 643 | 78 | ||
| KJ663722.1 | - | 3 - 4 | 565 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082929 | AK155650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082929 | NM_010389.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130916 | NM_010389.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229028 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130916 | NM_010389.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057931 | AK080011.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082929 | AK155650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082929 | NM_010389.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P679470 | AK053233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P679470 | NM_010389.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152821 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08764917 | - | - |
| Whole Mouse Genome G4122A | A_52_P679470 | AK053233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P679470 | NM_010389.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126717 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130916 | NM_010389.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08764917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152821 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444285 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422201_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440837_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444284 | - | - |
| Mouse Genome 430 2.0 Array | 1422201_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422201_at | - | - |
| Mouse Genome 430 2.0 Array | 1440837_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440837_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15002_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15002_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700006016.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006016.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006016.mm.1 | TC1700000625.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006019.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006019.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006019.mm.1 | TC1700000625.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006018.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006018.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006018.mm.1 | TC1700000625.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006014.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006014.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006014.mm.1 | TC1700000625.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006017.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006017.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006017.mm.1 | TC1700000625.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006020.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006020.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006020.mm.1 | TC1700000625.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006021.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006021.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006021.mm.1 | TC1700000625.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006013.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006013.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006013.mm.1 | TC1700000625.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006022.mm.1 | NM_010389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006022.mm.1 | NM_010389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006022.mm.1 | TC1700000625.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422201_at | - | - |
| Mouse Expression Set 430 | 1422201_PM_at | - | - |
| Mouse Expression Set 430 | 1440837_at | - | - |
| Mouse Expression Set 430 | 1440837_PM_at | - | - |
| Murine Genome U74v2 Set | 104429_PM_at | - | - |
| Murine Genome U74v2 Set | 104429_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422201_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422201_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422201_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422201_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440837_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440837_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 560648 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6980059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6980059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102690047 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980059 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2670690 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2502380 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2670690 | - | - |
| mouse-6 v1.1 (Illumina) | scl8544.1.1_308-S | - | - |
| mouse-6 v1.1 (Illumina) | scl50826.4.1_157-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980059 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102690047 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50826.4.1_157-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102690047 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980059 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2670690 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2670690 | - | - |

H2-Ob
Genomic Map