Assay Details
| Gene Symbol: Hira | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010435.2 | NP_034565.2 | 9 - 10 | 1674 | 58 | |
| GenBank mRNA | BC052856.1 | - | 9 - 10 | 1092 | 58 | |
| BC060181.1 | - | 9 - 10 | 1358 | 58 | ||
| X75295.1 | - | 10 - 11 | 1189 | 58 | ||
| X99712.1 | - | 9 - 10 | 1001 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | X99712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | AK168963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | BC060181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | AK135928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | NM_010435.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | X99712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | AK168963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | BC060181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | AK135928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P98232 | NM_010435.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387373 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221801 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 879614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147037 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489779 | NM_134243.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781331 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169758 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233881 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370142 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877595 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660856 | AF037208.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537330 | AK017011.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59410 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399042 | AK082634.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936698 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233260 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194206 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39469 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137843 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27925 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150816 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137134 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | X99712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | AK168963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | BC060181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | AK135928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | NM_010435.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | X99712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | AK168963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | BC060181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | AK135928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P98232 | NM_010435.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125395 | X75295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125395 | X92590.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125395 | X99712.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125395 | BC060181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125395 | NM_010435.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P49712 | X75295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P49712 | X92590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P49712 | X99712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P49712 | BC052856.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P49712 | BC060181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | X99712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | AK168963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | BC060181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | AK135928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | NM_010435.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | X99712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | AK168963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | BC060181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | AK135928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98232 | NM_010435.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125395 | X75295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125395 | X92590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125395 | X99712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125395 | BC060181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125395 | NM_010435.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118996 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137395 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12500589 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12928789 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12928783 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10434258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434263 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434277 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434264 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434268 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434276 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434275 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434266 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434280 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434274 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434279 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434273 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434265 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434261 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434272 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434262 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450050_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450049_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459103_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436241_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10434252 | - | - |
| Mouse Genome 430 2.0 Array | 1436241_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1436241_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1450049_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450049_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450050_at | - | - |
| Mouse Genome 430 2.0 Array | 1459103_at | - | - |
| Mouse Genome 430 2.0 Array | 1450050_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459103_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15260_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15260_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600002367.mm.1 | NM_010435.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002367.mm.1 | NM_010435.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002367.mm.1 | TC1600000253.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436241_PM_s_at | - | - |
| Mouse Expression Set 430 | 1450049_a_at | - | - |
| Mouse Expression Set 430 | 1450049_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450050_at | - | - |
| Mouse Expression Set 430 | 1450050_PM_at | - | - |
| Mouse Expression Set 430 | 1436241_s_at | - | - |
| Mouse Expression Set 430 | 1459103_PM_at | - | - |
| Mouse Expression Set 430 | 1459103_at | - | - |
| Murine Genome U74v2 Set | 103009_at | - | - |
| Murine Genome U74v2 Set | 102709_at | - | - |
| Murine Genome U74v2 Set | 102709_PM_at | - | - |
| Murine Genome U74v2 Set | 103009_PM_at | - | - |
| Murine Genome U74v2 Set | 171321_r_at | - | - |
| Murine Genome U74v2 Set | 171321_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450049_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450050_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450049_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436241_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436241_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450050_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450050_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450050_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450049_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436241_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436241_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450049_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459103_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459103_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 756314 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100630181 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270167 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480609 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270167 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2651880 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2717588 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2522378 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2717588 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2651880 | - | - |
| mouse-6 v1.1 (Illumina) | scl0015260.1_245-S | - | - |
| mouse-6 v1.1 (Illumina) | scl015260.11_119-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270167 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480609 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0015260.1_245-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl015260.11_119-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480609 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270167 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2651880 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239277 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239277 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2651880 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl015260.11_119-S | - | - |

Hira
Genomic Map