Assay Details
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011368.5 | NP_035498.2 | 1 - 2 | 166 | 52 | |
GenBank mRNA | AK049357.1 | - | 1 - 2 | 139 | 52 | |
BC036172.1 | - | 1 - 2 | 117 | 4988907 | 52 | |
U15784.1 | - | 1 - 2 | 56 | 52 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK049357.1,BC036172.1,NM_011368.5,U15784.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
