Assay Details
| Gene Symbol: Ercc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011729.2 | NP_035859.2 | 2 - 3 | 580 | 76 | |
| GenBank mRNA | AK038822.1 | - | 2 - 3 | 409 | 76 | |
| AK137563.1 | - | 2 - 3 | 412 | 76 | ||
| AK140825.1 | - | 2 - 3 | 409 | 76 | ||
| AK164205.1 | - | 2 - 3 | 398 | 76 | ||
| BC033413.1 | - | 2 - 3 | 300 | 76 | ||
| BC053034.1 | - | 2 - 3 | 389 | 76 | ||
| BC060659.1 | - | 2 - 3 | 379 | 76 | ||
| BC092235.1 | - | 2 - 3 | 371 | 76 | ||
| BC141070.1 | - | 2 - 3 | 410 | 9055682 | 76 | |
| D16306.1 | - | 2 - 3 | 275 | 76 | ||
| U40796.1 | - | 2 - 3 | 476 | 76 | ||
More Information
Set Membership: |
|

Ercc5
Genomic Map