Assay Details
| Gene Symbol: Eif2s3y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012011.1 | NP_036141.1 | 2 - 3 | 137 | 114 | |
| GenBank mRNA | AJ006584.1 | - | 2 - 3 | 137 | 114 | |
| AK009448.1 | - | 2 - 3 | 142 | 114 | ||
| AK153974.1 | - | 2 - 3 | 155 | 114 | ||
| BC031450.1 | - | 2 - 3 | 133 | 114 | ||
| BC043656.1 | - | 2 - 3 | 137 | 5368023 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011877 | BC043656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011877 | AJ006584.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011877 | AK009448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011877 | NM_012011.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189383 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 690402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528411 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 5267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10983 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60765 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011877 | BC043656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011877 | AJ006584.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011877 | AK009448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011877 | NM_012011.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32825353 | - | - |
| Whole Mouse Genome G4122A | A_51_P479715 | BC043656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479715 | AJ006584.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479715 | AK009448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479715 | NM_012011.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15232215 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15232210 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15229351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15229350 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15232218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32825480 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10608013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608012 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608011 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417210_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431642_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608001 | - | - |
| Mouse Genome 430 2.0 Array | 1417210_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431642_at | - | - |
| Mouse Genome 430 2.0 Array | 1457945_at | - | - |
| Mouse Genome 430 2.0 Array | 1431642_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457945_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417210_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26908_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26908_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0Y00000080.mm.1 | NM_012011.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0Y00000080.mm.1 | NM_012011.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0Y00000080.mm.1 | TC0Y00000006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0Y00000081.mm.1 | NM_012011.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0Y00000081.mm.1 | NM_012011.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0Y00000081.mm.1 | TC0Y00000006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0Y00000082.mm.1 | NM_012011.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0Y00000082.mm.1 | NM_012011.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0Y00000082.mm.1 | TC0Y00000006.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417210_PM_at | - | - |
| Mouse Expression Set 430 | 1417210_at | - | - |
| Mouse Expression Set 430 | 1431642_at | - | - |
| Mouse Expression Set 430 | 1431642_PM_at | - | - |
| Murine Genome U74v2 Set | 103674_f_at | - | - |
| Murine Genome U74v2 Set | 103674_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417210_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417210_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417210_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417210_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431642_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431642_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 931120 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6100707 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102510025 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006100707 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241001 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241001 | - | - |
| mouse-6 v1.1 (Illumina) | scl0026908.1_233-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6100707 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102510025 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0026908.1_233-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6100707 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102510025 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1241001 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215914 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241001 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0026908.1_233-S | - | - |

Eif2s3y
Genomic Map