Assay Details
| Gene Symbol: Epb4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013513.2 | NP_038541.1 | 2 - 3 | 357 | 68 | |
| GenBank mRNA | AK133896.1 | - | 2 - 3 | 371 | 68 | |
| AK137484.1 | - | 2 - 3 | 438 | 68 | ||
| AK143841.1 | - | 2 - 3 | 337 | 68 | ||
| AK158219.1 | - | 2 - 3 | 275 | 68 | ||
| AK158247.1 | - | 2 - 3 | 321 | 68 | ||
| AK158370.1 | - | 2 - 3 | 284 | 68 | ||
| AK165288.1 | - | 2 - 3 | 284 | 68 | ||
| AK172410.1 | - | 2 - 3 | 321 | 68 | ||
| BC053739.1 | - | 2 - 3 | 328 | 68 | ||
| BC066194.1 | - | 2 - 3 | 357 | 30429971 | 68 | |
| BC066806.1 | - | 2 - 3 | 357 | 30429961 | 68 | |
| L35933.1 | - | 2 - 3 | 336 | 68 | ||
| U04055.1 | - | 2 - 3 | 357 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epb4.2
Genomic Map