Assay Details
| Gene Symbol: H2-Ke6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013543.2 | NP_038571.2 | 5 - 6 | 567 | 88 | |
| GenBank mRNA | BC086927.1 | - | 5 - 6 | 567 | 6775958 | 88 |
| CX243612.1 | - | 5 - 6 | 536 | 88 | ||
| KJ650217.1 | - | 5 - 6 | 559 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
metabolic process
androgen metabolic process
estrogen metabolic process
oxidation-reduction process
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
3-hydroxyacyl-CoA dehydrogenase activity
aldo-keto reductase (NADP) activity
estradiol 17-beta-dehydrogenase activity
isocitrate dehydrogenase activity
mevaldate reductase activity
gluconate dehydrogenase activity
steroid dehydrogenase activity
oxidoreductase activity
epoxide dehydrogenase activity
5-exo-hydroxycamphor dehydrogenase activity
2-hydroxytetrahydrofuran dehydrogenase activity
acetoin dehydrogenase activity
phenylcoumaran benzylic ether reductase activity
D-xylose:NADP reductase activity
L-arabinose:NADP reductase activity
D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
steroid dehydrogenase activity, acting on the CH-CH group of donors
(R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
3-hydroxymenthone dehydrogenase activity
very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
dihydrotestosterone 17-beta-dehydrogenase activity
(R)-2-hydroxyisocaproate dehydrogenase activity
L-arabinose 1-dehydrogenase (NADP+) activity
L-xylulose reductase (NAD+) activity
testosterone dehydrogenase (NAD+) activity
3-ketoglucose-reductase activity
(R)-2-hydroxyglutarate dehydrogenase activity
D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity
lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
metabolic process
androgen metabolic process
estrogen metabolic process
oxidation-reduction process
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
3-hydroxyacyl-CoA dehydrogenase activity
aldo-keto reductase (NADP) activity
estradiol 17-beta-dehydrogenase activity
isocitrate dehydrogenase activity
mevaldate reductase activity
gluconate dehydrogenase activity
steroid dehydrogenase activity
oxidoreductase activity
epoxide dehydrogenase activity
5-exo-hydroxycamphor dehydrogenase activity
2-hydroxytetrahydrofuran dehydrogenase activity
acetoin dehydrogenase activity
phenylcoumaran benzylic ether reductase activity
D-xylose:NADP reductase activity
L-arabinose:NADP reductase activity
D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
steroid dehydrogenase activity, acting on the CH-CH group of donors
(R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
3-hydroxymenthone dehydrogenase activity
very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
dihydrotestosterone 17-beta-dehydrogenase activity
(R)-2-hydroxyisocaproate dehydrogenase activity
L-arabinose 1-dehydrogenase (NADP+) activity
L-xylulose reductase (NAD+) activity
testosterone dehydrogenase (NAD+) activity
3-ketoglucose-reductase activity
(R)-2-hydroxyglutarate dehydrogenase activity
D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151584 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151582 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073526 | BC086927.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073526 | NM_013543.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235680 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152697 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073526 | BC086927.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073526 | NM_013543.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P344407 | BC086927.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P344407 | CX243612.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P344407 | NM_013543.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151579 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152697 | - | - |
| Whole Mouse Genome G4122A | A_51_P344407 | BC086927.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344407 | CX243612.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344407 | NM_013543.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103515 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152697 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13118030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151584 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13133228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13118031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13133230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13150244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151571 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450113 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450111 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450107 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454987_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450103 | - | - |
| Mouse Genome 430 2.0 Array | 1454987_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1454987_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14979_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14979_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700017647.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017647.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017647.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017657.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017657.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017657.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017659.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017659.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017659.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017655.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017655.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017655.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017653.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017653.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017653.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017641.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017641.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017641.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017642.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017642.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017642.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017645.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017645.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017645.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017649.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017649.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017649.mm.1 | TC1700002810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017663.mm.1 | NM_013543.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017663.mm.1 | NM_013543.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017663.mm.1 | TC1700002810.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454987_PM_a_at | - | - |
| Mouse Expression Set 430 | 1454987_a_at | - | - |
| Murine Genome U74v2 Set | 102992_at | - | - |
| Murine Genome U74v2 Set | 102992_PM_at | - | - |
| Murine Genome U74v2 Set | 102991_PM_s_at | - | - |
| Murine Genome U74v2 Set | 102991_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454987_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454987_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454987_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454987_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 910217 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1940647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940647 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2653265 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2653265 | - | - |
| mouse-6 v1.1 (Illumina) | scl014979.1_326-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940647 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014979.1_326-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940647 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2653265 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2653265 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014979.1_326-S | - | - |

H2-Ke6
Genomic Map