Assay Details
| Gene Symbol: Tcirg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136091.2 | NP_001129563.1 | 11 - 12 | 1429 | 82 | |
| NM_001167784.1 | NP_001161256.1 | 11 - 12 | 1774 | 82 | ||
| NM_016921.3 | NP_058617.3 | 11 - 12 | 1470 | 82 | ||
| GenBank mRNA | AB022322.1 | - | 11 - 12 | 1336 | 82 | |
| AF218253.1 | - | 11 - 12 | 1408 | 82 | ||
| AF218254.1 | - | 5 - 6 | 1012 | 82 | ||
| AK004094.1 | - | 2 - 3 | 280 | 82 | ||
| AK155140.1 | - | 11 - 12 | 1775 | 82 | ||
| BC006761.1 | - | 11 - 12 | 1470 | 3257437 | 82 | |
| BC085234.1 | - | 11 - 12 | 1330 | 5322429 | 82 | |
More Information
Set Membership: |
|

Tcirg1
Genomic Map