Assay Details
| Gene Symbol: Eral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022313.2 | NP_071708.2 | 1 - 2 | 309 | 83 | |
| GenBank mRNA | AB049389.1 | - | 1 - 2 | 285 | 83 | |
| AF190730.1 | - | 1 - 2 | 277 | 83 | ||
| AF190731.1 | - | 1 - 2 | 294 | 83 | ||
| AK012155.1 | - | 1 - 2 | 299 | 83 | ||
| BC037603.1 | - | 1 - 2 | 294 | 83 | ||
| BC068271.1 | - | 1 - 2 | 304 | 30008388 | 83 | |
| BY158323.1 | - | 1 - 2 | 310 | 83 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF190730.1,AF190731.1 transcripts.
Set Membership: |
|

Eral1
Genomic Map